Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rdl
Entrez Gene ID
39054
Gene Name
Resistant to dieldrin
Gene Aliases
CG10537, CT29555, DmRDL, DmRdl, Dmel\CG10537, Dmel_CG10537, GABA, GABA-R, GABA[[A]], GABA[[A]] receptor, GABA[[A]]-R, GABA[[A]]R, GABAr, LCCH1, RDL, Rd1, gaba, rdl
Chromosome Location
Chr.3L: 9145838 - 9182149 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004244
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274686.1 NP_001261615.1 8 - 9 2690 70
NM_001274687.1 NP_001261616.1 8 - 9 2690 70
NM_001274688.1 NP_001261617.1 8 - 9 2690 70
NM_079267.4 NP_523991.2 7 - 8 1350 70
NM_168321.3 NP_729461.1 8 - 9 2690 70
NM_168322.3 NP_729462.2 8 - 9 2786 70

View Full Product Details