Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ciao1
Entrez Gene ID
36655
Gene Name
CG12797-PA
Gene Aliases
CG12797, CiaO1, Dmel\CG12797, Dmel_CG12797
Chromosome Location
Chr.2R: 14853863 - 14855048 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033972
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137152.4 NP_610996.1 1 - 1 921 90
GenBank mRNA AY061388.1 - 1 - 1 878 90

View Full Product Details