Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pod1
Entrez Gene ID
31620
Gene Name
pod1 coronin
Gene Aliases
BEST:LD37601, BcDNA:LD08951, CG4532, Dmel\CG4532, Dmel_CG4532, Dpod1, POD-1, Pod1, coronin, dmPod, dpod1, pod-1
Chromosome Location
Chr.X: 6803684 - 6813830 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029903
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103427.3 NP_001096897.1 8 - 9 2321 83
NM_132124.5 NP_572352.1 7 - 8 1817 83
NM_167091.3 NP_727125.1 7 - 8 1778 83
NM_167092.3 NP_727126.1 7 - 8 1777 83
NM_167093.2 NP_727127.1 7 - 8 1717 83
NM_167094.2 NP_727128.1 7 - 8 1786 83
GenBank mRNA BT003254.1 - 7 - 8 1764 83

View Full Product Details