Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hex-C
Entrez Gene ID
44008
Gene Name
Hexokinase C
Gene Aliases
CG8094, CT22069, DM2, Dmel\CG8094, Dmel_CG8094, Fk, HEX, HEX-3, HEX-C, HK, HK-C, Hex, Hex-3, Hex-c, HexC, Hk
Chromosome Location
Chr.2R: 15218837 - 15220329 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001187
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079935.4 NP_524674.1 1 - 1 1257 77
GenBank mRNA AF237469.1 - 1 - 1 1229 77
AY070657.1 - 1 - 1 1341 77
AY071719.1 - 1 - 1 1341 77

View Full Product Details