Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Pis
- Entrez Gene ID
-
32506
- Gene Name
- Phosphatidylinositol synthase
- Gene Aliases
-
CG9245, Dmel\CG9245, Dmel_CG9245, PIS, anon-WO02059370.38, dPIS, dPis, dpis
- Chromosome Location
-
Chr.X: 15701451 - 15703770 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0030670
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_167456.3 |
NP_727878.1 |
2 - 3 |
343 |
63 |
View Full Product Details