Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- wupA
- Entrez Gene ID
-
32794
- Gene Name
- wings up A
- Gene Aliases
-
CG7178, DROTROPONI, DmTpnI, Dmel\CG7178, Dmel_CG7178, HDP, HL-I, HL08042, HLI, Hdp, Tn I, TnI, TpnI, WupA, chrX:17947374..17947524, fliH, hdp, hdp-a, hdp2, wup-A, wupa
- Chromosome Location
-
Chr.X: 18105568 - 18116922 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0283471
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_167606.3 |
NP_728140.1 |
5 - 6 |
509 |
91 |
GenBank mRNA |
BT125791.1 |
- |
5 - 6 |
480 |
91 |
View Full Product Details