Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hfw
Entrez Gene ID
31120
Gene Name
halfway
Gene Aliases
CG3095, CT10406, Dmel\CG3095, Dmel_CG3095, EG:171E4.2, SWI, l(1)2Bf, l(1)nfw, swi
Chromosome Location
Chr.X: 1669570 - 1673608 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001189
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272206.1 NP_001259135.1 2 - 3 1285 66
NM_130577.4 NP_569933.2 2 - 3 1285 66
GenBank mRNA BT050442.1 - 2 - 3 1262 66

View Full Product Details