Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myo61F
Entrez Gene ID
38153
Gene Name
Myosin 61F
Gene Aliases
CG9155, Dm IB, DmIB, Dmel\CG9155, Dmel_CG9155, DroMIB, IB, MIB, MYO61F, MYOIB, Mhc2, Myo-IB, Myo1B, Myo1C, MyoIB, MyoIC, myo61F, myo61f, myoIC
Chromosome Location
Chr.3L: 1318480 - 1329617 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010246
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057586.4 NP_476934.2 4 - 5 792 73
NM_167870.2 NP_728594.2 4 - 5 869 73
NM_167871.2 NP_728595.1 4 - 5 877 73
NM_167872.2 NP_728596.1 4 - 5 1015 73

View Full Product Details