Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
shf
Entrez Gene ID
31617
Gene Name
shifted
Gene Aliases
CG3134, CG3135, CT10514, D-Wif, DmWIF, DmWif, Dmel\CG3135, Dmel_CG3135, Shf, shf-ov
Chromosome Location
Chr.X: 6771463 - 6779659 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003390
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298034.1 NP_001284963.1 3 - 4 913 77
NM_001298035.1 NP_001284964.1 3 - 4 913 77
NM_001298036.1 NP_001284965.1 3 - 4 913 77
NM_132121.3 NP_572349.1 3 - 4 913 77
GenBank mRNA AY051622.1 - 3 - 4 913 77

View Full Product Details