Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GIIIspla2
Entrez Gene ID
31747
Gene Name
Phospholipase A2 group III
Gene Aliases
BEST:HL07869, CG1583, Dmel\CG1583, Dmel_CG1583, GIIIsPLA2, Giiispla2
Chromosome Location
Chr.X: 8157628 - 8159348 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030013
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298079.1 NP_001285008.1 1 - 2 764 76
NM_132226.3 NP_572454.2 1 - 2 764 76

View Full Product Details