Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cbl
Entrez Gene ID
38961
Gene Name
Cbl proto-oncogene
Gene Aliases
CBL, CG7037, CG7043, D-Cbl, D-Cbl L, D-CblL, D-cbl, D-cblL, D-cblS, DCbl, Dcbl, Dmel\CG7037, Dmel_CG7037, Dv-Cbl, Dv-cbl, F165, anon-WO0118547.68, cbl, d-cbl, dCbl, v-Cbl
Chromosome Location
Chr.3L: 8424954 - 8432246 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020224
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139967.2 NP_648224.1 3 - 4 1444 76
NM_168276.2 NP_729382.1 3 - 4 1444 76
GenBank mRNA AF273749.1 - 3 - 4 1417 76
AJ223175.1 - 3 - 4 1416 76
AY118593.1 - 3 - 4 1109 76

View Full Product Details