Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rdl
Entrez Gene ID
39054
Gene Name
Resistant to dieldrin
Gene Aliases
CG10537, CT29555, DmRDL, DmRdl, Dmel\CG10537, Dmel_CG10537, GABA, GABA-R, GABA[[A]], GABA[[A]] receptor, GABA[[A]]-R, GABA[[A]]R, GABAr, LCCH1, RDL, Rd1, gaba, rdl
Chromosome Location
Chr.3L: 9145838 - 9182149 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004244
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274686.1 NP_001261615.1 4 - 5 1953 79
NM_168321.3 NP_729461.1 4 - 5 1953 79

View Full Product Details