Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
msk
Entrez Gene ID
44747
Gene Name
moleskin
Gene Aliases
61, CG7935, CIP-61, DIM-7, Dim-7, Dim7, DmRanBP7, Dmel\CG7935, Dmel_CG7935, Imp7, Imp7/8, Msk, S(ls)2, dim-7, dim7
Chromosome Location
Chr.3L: 8116179 - 8121579 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026252
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080041.3 NP_524780.1 2 - 3 590 74
GenBank mRNA AF132299.1 - 2 - 3 499 74
BT053682.1 - 2 - 3 591 74

View Full Product Details