Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
jub
Entrez Gene ID
32351
Gene Name
Ajuba LIM protein
Gene Aliases
Ajuba, CG11063, Dmel\CG11063, Dmel_CG11063, JUB, Jub, dJub, djub
Chromosome Location
Chr.X: 13826041 - 13830317 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030530
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201657.2 NP_001188586.1 4 - 5 2053 66
NM_001201658.2 NP_001188587.1 4 - 5 2068 66
NM_001298303.1 NP_001285232.1 4 - 5 2053 66
NM_132702.5 NP_572930.3 5 - 6 2083 66

View Full Product Details