Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bip1
Entrez Gene ID
38891
Gene Name
Bab interating protein 1
Gene Aliases
Bip1, CG7574, Dmel\CG7574, Dmel_CG7574, anon-WO0118547.246, bip-I
Chromosome Location
Chr.3L: 8054333 - 8058713 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026263
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169904.2 NP_001163375.1 2 - 3 364 58
NM_139912.5 NP_648169.2 3 - 4 717 58
GenBank mRNA BT089045.1 - 2 - 3 361 58

View Full Product Details