Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Myo61F
- Entrez Gene ID
-
38153
- Gene Name
- Myosin 61F
- Gene Aliases
-
CG9155, Dm IB, DmIB, Dmel\CG9155, Dmel_CG9155, DroMIB, IB, MIB, MYO61F, MYOIB, Mhc2, Myo-IB, Myo1B, Myo1C, MyoIB, MyoIC, myo61F, myo61f, myoIC
- Chromosome Location
-
Chr.3L: 1318480 - 1329617 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0010246
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057586.4 |
NP_476934.2 |
1 - 2 |
70 |
81 |
View Full Product Details