Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CycE
- Entrez Gene ID
-
34924
- Gene Name
- Cyclin E
- Gene Aliases
-
3938, BG:DS07108.3, CDI7, CG3938, CYCE, Cdi7, Cyc, Cyc E, Cyc-E, CycEI, CyclE, CyeE, D-CycE, DmCycE, DmcycE, DmcyclinE, Dmel\CG3938, Dmel_CG3938, br37, cdi7, cycE, cyclinE, cycline, dCycE, dm-cycE, fond, l(2)05206, l(2)35Dd, l(2)br37, l(2)k02514, l(2)k02602, l(2)k05007, l35Dd
- Chromosome Location
-
Chr.2L: 15727581 - 15748150 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0010382
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057612.5 |
NP_476960.1 |
1 - 2 |
788 |
70 |
View Full Product Details