Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- csul
- Entrez Gene ID
-
36809
- Gene Name
- capsuleen
- Gene Aliases
-
CG3730, Csul, DART5, Dart5, Dmel\CG3730, Dmel_CG3730, JBP1, PRMT5, dPRMT5, dart5, dprmt5
- Chromosome Location
-
Chr.2R: 16264652 - 16266959 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0015925
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057836.4 |
NP_477184.1 |
1 - 2 |
472 |
71 |
GenBank mRNA |
AY122153.1 |
- |
1 - 2 |
440 |
71 |
View Full Product Details