Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Hr3
- Entrez Gene ID
-
36073
- Gene Name
- Hormone receptor 3
- Gene Aliases
-
12.6, 2.4, CG11823, CG12208, CG33183, D, DHR3, Dhr3, DmHr3, DmRORalpha, Dmel\CG33183, Dmel_CG33183, HR3, Hr46, NR1F4, dHR3, dhr3, hr46, l(2)46CFi, l(2)k09242
- Chromosome Location
-
Chr.2R: 10203995 - 10237348 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000448
- Gene Symbol
- CG12912
- Entrez Gene ID
-
36075
- Gene Name
- CG12912-PA
- Gene Aliases
-
Dmel\CG12912, Dmel_CG12912
- Chromosome Location
-
Chr.2R: 10205454 - 10234607 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033497
View Full Product Details