Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hr3
Entrez Gene ID
36073
Gene Name
Hormone receptor 3
Gene Aliases
12.6, 2.4, CG11823, CG12208, CG33183, D, DHR3, Dhr3, DmHr3, DmRORalpha, Dmel\CG33183, Dmel_CG33183, HR3, Hr46, NR1F4, dHR3, dhr3, hr46, l(2)46CFi, l(2)k09242
Chromosome Location
Chr.2R: 10203995 - 10237348 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000448
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103786.2 NP_001097256.1 4 - 5 908 72
NM_001259307.3 NP_001246236.1 5 - 6 2740 72
NM_001273925.2 NP_001260854.1 5 - 6 2746 72
NM_001347757.1 NP_001334718.1 4 - 5 3141 72
NM_176121.3 NP_788301.1 4 - 5 3141 72
NM_176123.5 NP_788303.1 5 - 6 2746 72
GenBank mRNA BT133118.1 - 4 - 5 738 72

Gene Symbol
CG12912
Entrez Gene ID
36075
Gene Name
CG12912-PA
Gene Aliases
Dmel\CG12912, Dmel_CG12912
Chromosome Location
Chr.2R: 10205454 - 10234607 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033497
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299344.1 NP_001286273.1 4 - 5 3141 72

Gene Symbol
CG46321
Entrez Gene ID
30522410
Gene Name
CG46321-PA
Gene Aliases
Dmel\CG46321, Dmel_CG46321
Chromosome Location
Chr.2R: 10205454 - 10227957 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0284237
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001347803.1 NP_001334719.1 5 - 6 2746 72

View Full Product Details