Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
E2f1
Entrez Gene ID
42550
Gene Name
E2F transcription factor 1
Gene Aliases
CG6376, DRTF1/E2F, DmE2F-1, Dmel\CG6376, Dmel_CG6376, Dp, E(Sev-CycE)3A, E(var)3-93E, E(var)3-95E, E(var)93E, E2F, E2F-1, E2F1, E2f, Evar(3)164, dE2F, dE2F1, dE2f, dE2f1, de2f1, def21, drosE2F1, e2f, e2f1, l(3)07172, l(3)j3B1, l(3)j3C2, l(3)rM729
Chromosome Location
Chr.3R: 21620304 - 21660871 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011766
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300526.1 NP_001287455.1 5 - 6 2171 95
NM_079713.3 NP_524437.2 3 - 4 2495 95
NM_169961.3 NP_732646.1 5 - 6 1732 95
NM_169962.3 NP_732647.1 3 - 4 2296 95
GenBank mRNA BT016096.1 - 5 - 6 1705 95

View Full Product Details