Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
beta-Man
Entrez Gene ID
40524
Gene Name
beta-Mannosidase
Gene Aliases
CG12582, CG12583, Dmel\CG12582, Dmel_CG12582
Chromosome Location
Chr.3R: 4197206 - 4204572 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037215
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259970.2 NP_001246899.1 7 - 8 1757 62
NM_001275312.1 NP_001262241.1 6 - 7 1908 62
NM_001275313.1 NP_001262242.1 6 - 7 1870 62
NM_141179.4 NP_649436.2 6 - 7 1870 62
NM_164315.4 NP_730783.3 5 - 6 1942 62
GenBank mRNA BT044157.1 - 6 - 7 1769 62

View Full Product Details