Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
beta-Man
Entrez Gene ID
40524
Gene Name
beta-Mannosidase
Gene Aliases
CG12582, CG12583, Dmel\CG12582, Dmel_CG12582
Chromosome Location
Chr.3R: 4197206 - 4204572 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037215
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259970.2 NP_001246899.1 9 - 10 2563 77
NM_001275312.1 NP_001262241.1 8 - 9 2714 77
NM_141179.4 NP_649436.2 8 - 9 2676 77
NM_164315.4 NP_730783.3 7 - 8 2748 77
GenBank mRNA BT044157.1 - 8 - 9 2575 77

View Full Product Details