Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mia
Entrez Gene ID
40667
Gene Name
meiosis I arrest
Gene Aliases
CG10390, Dmel\CG10390, Dmel_CG10390, Mia, TAF, TAF60-2, TAF6L, TAF[[II]], TAF[[II]]60-2, TFIID, Taf60-2, Taf6L, dTAF6L, dTAF[[II]]60, dTAF[[II]]60-2, dmTAF6b, ms(3)570, taf6L
Chromosome Location
Chr.3R: 5548115 - 5551168 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0014342
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144538.2 NP_001138010.1 1 - 2 810 69
NM_001300252.1 NP_001287181.1 1 - 2 1346 69
NM_141290.6 NP_649547.3 1 - 2 810 69

View Full Product Details