Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mura
Entrez Gene ID
41145
Gene Name
murashka
Gene Aliases
CG9381, CG9382, Dmel\CG9381, Dmel_CG9381, Mura
Chromosome Location
Chr.3R: 9535625 - 9551806 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037705
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141645.4 NP_649902.2 2 - 3 616 172
NM_169291.2 NP_731366.1 3 - 4 649 172
GenBank mRNA AY094811.1 - 2 - 3 591 172

View Full Product Details