Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp6d5
Entrez Gene ID
41706
Gene Name
CG3050-PA
Gene Aliases
6d5, CG3050, CYP6d5, Cyp6D5, Dmel\CG3050, Dmel_CG3050, cyp6d5
Chromosome Location
Chr.3R: 14029228 - 14031483 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038194
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_142070.3 NP_650327.1 3 - 4 1174 72
GenBank mRNA AY118299.1 - 3 - 4 1159 72

View Full Product Details