Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cad89D
- Entrez Gene ID
-
42006
- Gene Name
- Cadherin 89D
- Gene Aliases
-
CG14900, CT34724, DCad89D, Dmel\CG14900, Dmel_CG14900
- Chromosome Location
-
Chr.3R: 16484566 - 16495422 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038439
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142297.4 |
NP_650554.3 |
5 - 6 |
2377 |
75 |
View Full Product Details