Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hel89B
Entrez Gene ID
41943
Gene Name
Helicase 89B
Gene Aliases
89B, BTAF1, CG4261, Dmel\CG4261, Dmel_CG4261, HEL89B, Helicase89B, Mot1
Chromosome Location
Chr.3R: 15973298 - 15983451 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0022787
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300422.1 NP_001287351.1 3 - 4 1084 64
NM_169693.2 NP_732097.2 3 - 4 1511 64

View Full Product Details