Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GluRIIE
Entrez Gene ID
318623
Gene Name
Glutamate receptor IIE
Gene Aliases
CG31201, DmelGluRIIE, Dmel\CG31201, Dmel_CG31201, GluR-IIE
Chromosome Location
Chr.3R: 20644154 - 20648716 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0051201
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043268.2 NP_001036733.1 3 - 4 620 96
GenBank mRNA BT056325.1 - 3 - 4 618 96

View Full Product Details