Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cenpp
Entrez Gene ID
445161
Gene Name
centromere protein P
Gene Aliases
fb81h12, si:ch211-103f16.3, wu:fb81h12, zgc:101125
Chromosome Location
Chr.22: 10441004 - 10513965 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003555.3 NP_001003555.2 8 - 9 815 116
GenBank mRNA BC078335.1 - 8 - 9 789 116

View Full Product Details