Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oclnb
Entrez Gene ID
494075
Gene Name
occludin b
Gene Aliases
wu:fk83f10, zgc:101820
Chromosome Location
Chr.5: 26212578 - 26220871 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008618.1 NP_001008618.1 4 - 5 888 90
XM_005171844.4 XP_005171901.1 3 - 4 976 90
GenBank mRNA BC086743.1 - 4 - 5 888 90
BC164682.1 - 3 - 4 742 90

View Full Product Details