Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
il19l
Entrez Gene ID
100004224
Gene Name
interleukin 19 like
Gene Aliases
-
Chromosome Location
Chr.11: 21096339 - 21099783 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082955.1 NP_001076424.1 1 - 2 185 68
GenBank mRNA AB219579.1 - 1 - 2 185 68

View Full Product Details