Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
il19l
Entrez Gene ID
100004224
Gene Name
interleukin 19 like
Gene Aliases
-
Chromosome Location
Chr.11: 21096339 - 21099783 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082955.1 NP_001076424.1 4 - 5 473 80
GenBank mRNA AB219579.1 - 4 - 5 473 80

View Full Product Details