Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tbx2b
Entrez Gene ID
30253
Gene Name
T-box transcription factor 2b
Gene Aliases
hm:zeh1581, tbx-c, tbx2, tbxc, zeh1581
Chromosome Location
Chr.15: 76306 - 83248 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131051.1 NP_571126.1 5 - 6 1444 73
XM_017359202.2 XP_017214691.1 3 - 4 388 73
GenBank mRNA AF136946.1 - 5 - 6 997 73
AF179405.1 - 5 - 6 1444 73
BC051603.1 - 5 - 6 1440 73

View Full Product Details