Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
egr1
Entrez Gene ID
30498
Gene Name
early growth response 1
Gene Aliases
krox-24, krox24, wu:fj64b05, wu:fq25f01
Chromosome Location
Chr.14: 181169 - 183410 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131248.1 NP_571323.1 1 - 2 210 119
GenBank mRNA BC163873.1 - 1 - 2 246 119

View Full Product Details