Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyb5d1
Entrez Gene ID
795592
Gene Name
cytochrome b5 domain containing 1
Gene Aliases
zgc:112008
Chromosome Location
Chr.7: 20771752 - 20778572 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017684.2 NP_001017684.1 3 - 4 501 114
GenBank mRNA BC093157.1 - 3 - 4 504 114

View Full Product Details