Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mkrn1
Entrez Gene ID
170782
Gene Name
makorin, ring finger protein, 1
Gene Aliases
fl25h08, wu:fl25h08, zgc:110403
Chromosome Location
Chr.4: 12292285 - 12307039 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152978.1 NP_694510.1 5 - 6 866 84
GenBank mRNA AF277173.2 - 5 - 6 837 84
BC095243.1 - 5 - 6 866 84
BC164736.1 - 5 - 6 887 84

View Full Product Details