Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tbx20
Entrez Gene ID
57936
Gene Name
T-box transcription factor 20
Gene Aliases
ZF-TBX20, cb826, hrt, whz
Chromosome Location
Chr.16: 42505025 - 42523910 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131506.2 NP_571581.2 6 - 7 1055 66
GenBank mRNA AF239664.1 - 6 - 7 1057 66
AF253325.1 - 6 - 7 1070 66
BC163475.1 - 6 - 7 1046 66

View Full Product Details