Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aar2
Entrez Gene ID
431768
Gene Name
AAR2 splicing factor
Gene Aliases
im:7153138, zgc:92018
Chromosome Location
Chr.23: 4216193 - 4225606 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002221.1 NP_001002221.1 3 - 4 1012 66
XM_005161924.4 XP_005161981.1 4 - 5 1122 66
GenBank mRNA BC072724.1 - 3 - 4 1012 66

View Full Product Details