Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lin7b
Entrez Gene ID
553768
Gene Name
lin-7 homolog B (C. elegans)
Gene Aliases
fj41b11, fj63g03, wu:fj41b11, wu:fj63g03, zgc:109997
Chromosome Location
Chr.3: 31933925 - 31944709 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020733.2 NP_001018569.2 1 - 2 209 71
GenBank mRNA AY584746.1 - 1 - 2 209 71
BC095116.1 - 1 - 2 241 71
BC165891.1 - 1 - 2 75 71

View Full Product Details