Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mettl16
Entrez Gene ID
445217
Gene Name
methyltransferase like 16
Gene Aliases
fi09d09, im:7148386, mett10d, wu:fi09d09, zgc:100981, zgc:158576
Chromosome Location
Chr.15: 25214171 - 25268619 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003611.1 NP_001003611.1 5 - 6 667 62
GenBank mRNA BC078220.1 - 5 - 6 667 62

View Full Product Details