Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mettl16
Entrez Gene ID
445217
Gene Name
methyltransferase like 16
Gene Aliases
fi09d09, im:7148386, mett10d, wu:fi09d09, zgc:100981, zgc:158576
Chromosome Location
Chr.15: 25214171 - 25268619 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003611.1 NP_001003611.1 8 - 9 971 140
GenBank mRNA BC078220.1 - 8 - 9 971 140

View Full Product Details