Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
adrm1
Entrez Gene ID
796450
Gene Name
adhesion regulating molecule 1
Gene Aliases
adrm1a, adrm1b, fa12d09, wu:fa10g10, wu:fa12d09, zgc:56107, zgc:77256
Chromosome Location
Chr.23: 9560965 - 9567306 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201013.3 NP_957307.3 8 - 9 1171 71
XM_005161962.4 XP_005162019.1 8 - 9 1187 71
XM_021469854.1 XP_021325529.1 8 - 9 1184 71
GenBank mRNA BC045915.1 - 7 - 8 977 71
BC066391.1 - 8 - 9 1150 71
BC154825.1 - 8 - 9 1113 71
EH561147.1 - 2 - 3 343 71

View Full Product Details