Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cul1a
Entrez Gene ID
323883
Gene Name
cullin 1a
Gene Aliases
Cul1, wu:fc13a10, zgc:55747, zgc:77500
Chromosome Location
Chr.2: 326373 - 369093 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199659.2 NP_955953.2 19 - 20 2375 97
XM_021478841.1 XP_021334516.1 19 - 20 2403 97
GenBank mRNA BC045445.1 - 19 - 20 2359 97
BC066480.1 - 19 - 20 2375 97
BC165664.1 - 18 - 19 2074 97

View Full Product Details