Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cul1a
Entrez Gene ID
323883
Gene Name
cullin 1a
Gene Aliases
Cul1, wu:fc13a10, zgc:55747, zgc:77500
Chromosome Location
Chr.2: 326373 - 369093 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199659.2 NP_955953.2 7 - 8 1130 90
XM_021478841.1 XP_021334516.1 7 - 8 1158 90
GenBank mRNA BC066480.1 - 7 - 8 1130 90

View Full Product Details