Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zdhhc4
Entrez Gene ID
561817
Gene Name
zinc finger DHHC-type palmitoyltransferase 4
Gene Aliases
fa97b04, wu:fa97b04, zgc:64155
Chromosome Location
Chr.12: 537941 - 544467 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200049.2 NP_956343.2 5 - 6 574 60
GenBank mRNA BC053285.1 - 5 - 6 571 60
BC152669.1 - 4 - 5 502 60
BC154785.1 - 6 - 7 880 60

View Full Product Details