Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eomesa
Entrez Gene ID
64603
Gene Name
eomesodermin homolog a
Gene Aliases
eom, eomes
Chromosome Location
Chr.19: 860825 - 868299 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131679.3 NP_571754.3 4 - 5 1281 87
XM_005159319.4 XP_005159376.1 5 - 6 1729 87
XM_005159321.4 XP_005159378.1 5 - 6 1615 87
GenBank mRNA AF287007.2 - 5 - 6 1185 87
AF329830.1 - 4 - 5 1243 87
BC067719.1 - 4 - 5 1281 87
BC154205.1 - 5 - 6 1504 87

View Full Product Details