Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
eomesa
Entrez Gene ID
64603
Gene Name
eomesodermin homolog a
Gene Aliases
eom, eomes
Chromosome Location
Chr.19: 860825 - 868299 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131679.3 NP_571754.3 5 - 6 1344 63
XM_005159319.4 XP_005159376.1 6 - 7 1792 63
XM_005159321.4 XP_005159378.1 6 - 7 1678 63
GenBank mRNA AF287007.2 - 6 - 7 1248 63
AF329830.1 - 5 - 6 1306 63
BC067719.1 - 5 - 6 1344 63
BC154205.1 - 6 - 7 1567 63

View Full Product Details