Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dvl1b
Entrez Gene ID
560032
Gene Name
dishevelled segment polarity protein 1b
Gene Aliases
dvl1, dvl2l
Chromosome Location
Chr.11: 24360582 - 24428628 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044917.1 NP_001038382.1 9 - 10 966 68
XM_005169297.4 XP_005169354.1 9 - 10 1384 68
XM_005169298.4 XP_005169355.1 9 - 10 1384 68
XM_005169299.4 XP_005169356.1 7 - 8 1202 68
XM_005169301.4 XP_005169358.1 9 - 10 1384 68
GenBank mRNA BC171461.1 - 9 - 10 1075 68

View Full Product Details