Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
thbs3b
Entrez Gene ID
571317
Gene Name
thrombospondin 3b
Gene Aliases
si:dkey-81h8.2
Chromosome Location
Chr.19: 24416183 - 24443736 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045214.1 NP_001038679.1 12 - 13 1361 79
XM_005170062.4 XP_005170119.1 12 - 13 1772 79
GenBank mRNA BC163250.1 - 11 - 12 1319 79

View Full Product Details